The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005269675.1 | XP_005269732.1 | 1 - 2 | 410 | 88 |
GenBank mRNA | FN908146.1 | - | 1 - 2 | 112 | 88 |
FR686760.1 | - | 1 - 2 | 105 | 88 |