The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000338.2 | NP_000329.2 | 3 - 4 | 767 | 66 |
NM_001184832.1 | NP_001171761.1 | 3 - 4 | 767 | 66 | |
XM_005254606.1 | XP_005254663.1 | 3 - 4 | 767 | 66 | |
GenBank mRNA | AK295375.1 | - | 2 - 3 | 220 | 66 |
AK298312.1 | - | 3 - 4 | 766 | 66 | |
BC040138.2 | - | 2 - 3 | 737 | 66 | |
EF559316.2 | - | 2 - 3 | 565 | 66 | |
U58130.1 | - | 2 - 3 | 570 | 66 |