Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GZMK
Entrez Gene ID
3003
Gene Name
granzyme K
Gene Aliases
TRYP2
Chromosome Location
Chr.5: 55024279 - 55034132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002104.2 NP_002095.1 2 - 3 256 74
GenBank mRNA AK312074.1 - 2 - 3 279 74
BC035802.1 - 2 - 3 256 74
U26174.1 - 2 - 3 252 74
U35237.1 - 2 - 3 212 74

View Full Product Details