The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000039.2 | NP_000030.1 | 1 - 2 | 352 | 70 |
NM_001318021.1 | NP_001304950.1 | 1 - 2 | 352 | 70 | |
GenBank mRNA | BC110286.1 | - | 1 - 2 | 47 | 70 |
CD558050.1 | - | 1 - 2 | 352 | 70 |