The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000204.4 | NP_000195.2 | 9 - 10 | 1297 | 79 |
NM_001318057.1 | NP_001304986.1 | 10 - 11 | 1321 | 79 | |
XM_005262976.1 | XP_005263033.1 | 8 - 9 | 1276 | 79 | |
XM_006714209.1 | XP_006714272.1 | 10 - 11 | 1318 | 79 | |
XM_006714210.3 | XP_006714273.1 | 10 - 11 | 1321 | 79 | |
XM_011531920.1 | XP_011530222.1 | 10 - 11 | 1321 | 79 | |
XM_017008164.1 | XP_016863653.1 | 9 - 10 | 1297 | 79 | |
XM_017008165.1 | XP_016863654.1 | 8 - 9 | 1276 | 79 | |
XM_017008166.1 | XP_016863655.1 | 9 - 10 | 1297 | 79 | |
GenBank mRNA | AK122686.1 | - | 6 - 7 | 1464 | 79 |
AK290625.1 | - | 9 - 10 | 1070 | 79 | |
AK299232.1 | - | 10 - 11 | 1147 | 79 | |
J02770.1 | - | 9 - 10 | 1057 | 79 | |
KU159429.1 | - | 9 - 10 | 1277 | 79 | |
Y00318.1 | - | 9 - 10 | 1072 | 79 |