The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001463.3 | NP_001454.2 | 5 - 6 | 1078 | 73 |
GenBank mRNA | AK130009.1 | - | 6 - 7 | 730 | 73 |
AK312741.1 | - | 5 - 6 | 957 | 73 | |
BC027855.1 | - | 5 - 6 | 930 | 73 | |
BT019883.1 | - | 5 - 6 | 860 | 73 | |
U24163.1 | - | 5 - 6 | 1068 | 73 | |
U68057.1 | - | 5 - 6 | 920 | 73 | |
U91903.1 | - | 5 - 6 | 929 | 73 |