The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015880.1 | NP_001015880.1 | 3 - 4 | 638 | 77 |
NM_004670.3 | NP_004661.2 | 3 - 4 | 638 | 77 | |
GenBank mRNA | AF074331.1 | - | 3 - 4 | 438 | 77 |
AF091242.1 | - | 3 - 4 | 455 | 77 | |
AF150754.2 | - | 3 - 4 | 455 | 77 | |
AF173365.1 | - | 3 - 4 | 423 | 77 | |
AF313907.1 | - | 3 - 4 | 375 | 77 | |
AK301453.1 | - | 3 - 4 | 425 | 77 | |
AK309262.1 | - | 3 - 4 | 447 | 77 | |
AK312556.1 | - | 3 - 4 | 479 | 77 | |
BC009894.2 | - | 3 - 4 | 486 | 77 | |
BQ221395.1 | - | 3 - 4 | 646 | 77 | |
CB960618.1 | - | 3 - 4 | 517 | 77 |