The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172673.1 | NP_001166144.1 | 1 - 2 | 294 | 63 |
NM_021938.3 | NP_068757.2 | 1 - 2 | 295 | 63 | |
XM_006722832.1 | XP_006722895.1 | 1 - 2 | 322 | 63 | |
XM_006722833.1 | XP_006722896.1 | 1 - 2 | 322 | 63 | |
GenBank mRNA | AB209067.1 | - | 1 - 2 | 294 | 63 |
AF329266.1 | - | 1 - 2 | 258 | 63 | |
BC028101.1 | - | 1 - 2 | 295 | 63 |