The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134707.1 | NP_001128179.1 | 20 - 21 | 2767 | 66 |
NM_007101.3 | NP_009032.2 | 20 - 21 | 2926 | 66 | |
GenBank mRNA | AF095735.2 | - | 20 - 21 | 2891 | 66 |
AF162428.1 | - | 19 - 20 | 2663 | 66 | |
AK093794.1 | - | 8 - 9 | 1817 | 66 | |
AK223402.1 | - | 20 - 21 | 2813 | 66 | |
AK291211.1 | - | 20 - 21 | 2761 | 66 | |
AK298348.1 | - | 20 - 21 | 2810 | 66 | |
AK316494.1 | - | 19 - 20 | 2565 | 66 | |
BC033217.1 | - | - | 1564 | 66 | |
BC041574.1 | - | 13 - 14 | 1573 | 66 | |
BC136363.1 | - | 20 - 21 | 2744 | 66 | |
BC136364.1 | - | 20 - 21 | 2744 | 66 | |
BC144035.1 | - | 20 - 21 | 2762 | 66 |