The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001053.3 | NP_001044.1 | 1 - 1 | 1111 | 157 |
NM_001172560.1 | NP_001166031.1 | 2 - 2 | 1094 | 157 | |
GenBank mRNA | AK308381.1 | - | 1 - 1 | 1111 | 156 |