Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KLHL9
- Entrez Gene ID
-
55958
- Gene Name
- kelch like family member 9
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 21331019 - 21335430 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC052776.1 |
- |
1 - 1 |
968 |
141 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
interferon alpha 5 |
IFNA5 |
IFN-alpha-5, IFN-alphaG, INA5, INFA5, leIF G |
-24.4kb
|
Sense |
Chr. - 21304614 - 21305313 on Build GRCh38 |
Downstream |
interferon alpha 6 |
IFNA6 |
IFN-alphaK |
18.3kb
|
Sense |
Chr. - 21350318 - 21350887 on Build GRCh38 |
View Full Product Details