The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017797.3 | NP_060267.2 | 4 - 5 | 790 | 75 |
XM_005259593.1 | XP_005259650.1 | 4 - 5 | 555 | 75 | |
XM_011528127.1 | XP_011526429.1 | 2 - 3 | 156 | 75 | |
GenBank mRNA | AF355797.1 | - | 4 - 5 | 790 | 75 |
AK000393.1 | - | 1 - 2 | 584 | 75 | |
AK027481.1 | - | 2 - 3 | 152 | 75 | |
AK225831.1 | - | 4 - 5 | 789 | 75 | |
AY007116.1 | - | 4 - 5 | 512 | 75 | |
BC000564.2 | - | 1 - 2 | 80 | 75 | |
BC008035.2 | - | 1 - 2 | 74 | 75 | |
BC069199.1 | - | 2 - 3 | 216 | 75 | |
BC094820.1 | - | 4 - 5 | 524 | 75 |