Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTBD2
Entrez Gene ID
55643
Gene Name
BTB domain containing 2
Gene Aliases
-
Chromosome Location
Chr.19: 1985438 - 2015703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017797.3 NP_060267.2 8 - 9 1417 69
XM_005259593.1 XP_005259650.1 8 - 9 1182 69
XM_011528127.1 XP_011526429.1 6 - 7 783 69
GenBank mRNA AF355797.1 - 8 - 9 1417 69
AK000393.1 - 5 - 6 1211 69
AK027481.1 - 6 - 7 779 69
AK056818.1 - 2 - 3 1552 69
AK225831.1 - 8 - 9 1416 69
AL360275.1 - 5 - 6 648 69
AY007116.1 - 8 - 9 1139 69
BC000564.2 - 5 - 6 707 69
BC008035.2 - 5 - 6 701 69
BC053986.1 - 4 - 5 602 69
BC069199.1 - 6 - 7 843 69
BC094820.1 - 8 - 9 1151 69
CR457230.1 - 4 - 5 454 69

View Full Product Details