The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016239.3 | NP_057323.3 | 37 - 38 | 7731 | 58 |
XM_017024714.1 | XP_016880203.1 | 34 - 35 | 8031 | 58 | |
XM_017024715.1 | XP_016880204.1 | 35 - 36 | 8094 | 58 |