Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIP12
Entrez Gene ID
9320
Gene Name
thyroid hormone receptor interactor 12
Gene Aliases
TRIP-12, ULF
Chromosome Location
Chr.2: 229763837 - 229923234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284214.1 NP_001271143.1 20 - 21 3097 129
NM_001284215.1 NP_001271144.1 18 - 19 2922 129
NM_001284216.1 NP_001271145.1 17 - 18 2143 129
NM_004238.2 NP_004229.1 19 - 20 2953 129
XM_005246954.4 XP_005247011.1 20 - 21 3205 129
XM_005246955.4 XP_005247012.1 20 - 21 3204 129
XM_005246956.4 XP_005247013.1 20 - 21 3181 129
XM_005246957.4 XP_005247014.1 20 - 21 3171 129
XM_005246958.4 XP_005247015.1 20 - 21 3124 129
XM_005246960.4 XP_005247017.1 21 - 22 3118 129
XM_005246961.3 XP_005247018.1 19 - 20 3055 129
XM_005246962.4 XP_005247019.1 19 - 20 3052 129
XM_006712852.3 XP_006712915.1 19 - 20 3008 129
XM_011512180.2 XP_011510482.1 20 - 21 3158 129
XM_011512181.2 XP_011510483.1 20 - 21 3156 129
XM_011512183.2 XP_011510485.1 19 - 20 3012 129
XM_017005272.1 XP_016860761.1 21 - 22 3264 129
XM_017005273.1 XP_016860762.1 20 - 21 3131 129
XM_017005274.1 XP_016860763.1 20 - 21 3079 129
XM_017005275.1 XP_016860764.1 20 - 21 3134 129
XM_017005276.1 XP_016860765.1 20 - 21 3139 129
XM_017005277.1 XP_016860766.1 21 - 22 3279 129
XM_017005278.1 XP_016860767.1 20 - 21 3094 129
XM_017005279.1 XP_016860768.1 20 - 21 3166 129
XM_017005280.1 XP_016860769.1 20 - 21 3155 129
XM_017005281.1 XP_016860770.1 20 - 21 3074 129
XM_017005282.1 XP_016860771.1 21 - 22 3117 129
XM_017005283.1 XP_016860772.1 20 - 21 3121 129
XM_017005284.1 XP_016860773.1 21 - 22 3115 129
XM_017005285.1 XP_016860774.1 19 - 20 3042 129
XM_017005286.1 XP_016860775.1 19 - 20 3074 129
XM_017005287.1 XP_016860776.1 19 - 20 3055 129
XM_017005288.1 XP_016860777.1 19 - 20 3017 129
XM_017005289.1 XP_016860778.1 21 - 22 3037 129
XM_017005290.1 XP_016860779.1 21 - 22 3034 129
XM_017005291.1 XP_016860780.1 20 - 21 2968 129
XM_017005292.1 XP_016860781.1 19 - 20 2971 129
XM_017005293.1 XP_016860782.1 19 - 20 2971 129
GenBank mRNA BC113891.1 - 17 - 18 2037 129
BC114556.1 - 20 - 21 2991 129
D28476.1 - 19 - 20 2883 129
EU489742.1 - 18 - 19 2876 129

View Full Product Details