Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TRIP12
- Entrez Gene ID
-
9320
- Gene Name
- thyroid hormone receptor interactor 12
- Gene Aliases
-
TRIP-12, ULF
- Chromosome Location
-
Chr.2: 229763837 - 229923234 on Build GRCh38
View Full Product Details