Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIP12
Entrez Gene ID
9320
Gene Name
thyroid hormone receptor interactor 12
Gene Aliases
TRIP-12, ULF
Chromosome Location
Chr.2: 229763837 - 229923234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284214.1 NP_001271143.1 39 - 40 5905 73
NM_001284215.1 NP_001271144.1 37 - 38 5730 73
NM_001284216.1 NP_001271145.1 36 - 37 4951 73
NM_004238.2 NP_004229.1 38 - 39 5761 73
XM_005246954.4 XP_005247011.1 39 - 40 6013 73
XM_005246955.4 XP_005247012.1 39 - 40 6012 73
XM_005246956.4 XP_005247013.1 39 - 40 5989 73
XM_005246957.4 XP_005247014.1 39 - 40 5964 73
XM_005246958.4 XP_005247015.1 39 - 40 5932 73
XM_005246960.4 XP_005247017.1 40 - 41 5926 73
XM_005246961.3 XP_005247018.1 38 - 39 5863 73
XM_005246962.4 XP_005247019.1 38 - 39 5860 73
XM_006712852.3 XP_006712915.1 38 - 39 5816 73
XM_011512180.2 XP_011510482.1 39 - 40 5966 73
XM_011512181.2 XP_011510483.1 39 - 40 5964 73
XM_011512183.2 XP_011510485.1 38 - 39 5820 73
XM_017005272.1 XP_016860761.1 40 - 41 6072 73
XM_017005273.1 XP_016860762.1 39 - 40 5939 73
XM_017005274.1 XP_016860763.1 39 - 40 5887 73
XM_017005275.1 XP_016860764.1 39 - 40 5942 73
XM_017005276.1 XP_016860765.1 39 - 40 5947 73
XM_017005277.1 XP_016860766.1 40 - 41 6087 73
XM_017005278.1 XP_016860767.1 39 - 40 5902 73
XM_017005279.1 XP_016860768.1 39 - 40 5959 73
XM_017005280.1 XP_016860769.1 39 - 40 5948 73
XM_017005281.1 XP_016860770.1 39 - 40 5882 73
XM_017005282.1 XP_016860771.1 40 - 41 5925 73
XM_017005283.1 XP_016860772.1 39 - 40 5914 73
XM_017005284.1 XP_016860773.1 40 - 41 5908 73
XM_017005285.1 XP_016860774.1 38 - 39 5850 73
XM_017005286.1 XP_016860775.1 38 - 39 5882 73
XM_017005287.1 XP_016860776.1 38 - 39 5848 73
XM_017005288.1 XP_016860777.1 38 - 39 5810 73
XM_017005289.1 XP_016860778.1 40 - 41 5845 73
XM_017005290.1 XP_016860779.1 40 - 41 5842 73
XM_017005291.1 XP_016860780.1 39 - 40 5776 73
XM_017005292.1 XP_016860781.1 38 - 39 5764 73
XM_017005293.1 XP_016860782.1 38 - 39 5779 73
GenBank mRNA BC113891.1 - 36 - 37 4845 73
BC114556.1 - 39 - 40 5799 73
D28476.1 - 38 - 39 5691 73
EU489742.1 - 37 - 38 5684 73
L40383.1 - 2 - 3 182 73

View Full Product Details