The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002862.2 | NP_001002862.1 | 5 - 6 | 549 | 73 |
NM_001135751.1 | NP_001129223.1 | 5 - 6 | 549 | 73 | |
NM_198440.3 | NP_940842.2 | 5 - 6 | 549 | 73 | |
XM_011530505.2 | XP_011528807.1 | 5 - 6 | 659 | 73 | |
XM_011530506.2 | XP_011528808.1 | 5 - 6 | 657 | 73 | |
XM_017029078.1 | XP_016884567.1 | 5 - 6 | 796 | 73 | |
XM_017029079.1 | XP_016884568.1 | 5 - 6 | 954 | 73 | |
XM_017029080.1 | XP_016884569.1 | 5 - 6 | 912 | 73 | |
XM_017029082.1 | XP_016884571.1 | 4 - 5 | 554 | 73 | |
GenBank mRNA | AK125830.1 | - | 4 - 5 | 750 | 73 |
AL389876.1 | - | 5 - 6 | 549 | 73 | |
AW135031.1 | - | 2 - 3 | 252 | 73 | |
BC057830.1 | - | 5 - 6 | 542 | 73 | |
BM792257.1 | - | 3 - 4 | 356 | 73 | |
CR456372.1 | - | 5 - 6 | 543 | 73 |