Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSH2D
Entrez Gene ID
84941
Gene Name
hematopoietic SH2 domain containing
Gene Aliases
ALX, HSH2
Chromosome Location
Chr.19: 16134028 - 16158575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC016826.1 - 1 - 2 49 59

View Full Product Details