The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260.2 | NP_001251.1 | 5 - 6 | 1054 | 128 |
NM_001318368.1 | NP_001305297.1 | 5 - 6 | 1054 | 128 | |
XM_005266216.4 | XP_005266273.1 | 5 - 6 | 412 | 128 | |
XM_017020317.1 | XP_016875806.1 | 4 - 5 | 410 | 128 | |
GenBank mRNA | AK296875.1 | - | 4 - 5 | 397 | 128 |
BC069634.1 | - | 5 - 6 | 539 | 128 | |
BC104492.1 | - | 5 - 6 | 542 | 128 | |
BC105950.1 | - | 5 - 6 | 542 | 128 | |
BC107601.1 | - | 5 - 6 | 541 | 128 | |
X85753.1 | - | 5 - 6 | 542 | 128 |