Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRG2
Entrez Gene ID
9542
Gene Name
neuregulin 2
Gene Aliases
DON1, HRG2, NTAK
Chromosome Location
Chr.5: 139846779 - 140043675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013982.2 NP_053585.1 5 - 6 1425 65

View Full Product Details