The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162429.2 | NP_001155901.1 | 1 - 2 | 364 | 159 |
NM_001256192.1 | NP_001243121.1 | 1 - 2 | 364 | 159 | |
NM_013374.5 | NP_037506.2 | 1 - 2 | 364 | 159 | |
GenBank mRNA | AF151793.1 | - | 1 - 2 | 332 | 159 |
AF250394.1 | - | 1 - 2 | 296 | 159 | |
AF349951.1 | - | 1 - 2 | 337 | 159 | |
BC020066.1 | - | 1 - 2 | 314 | 159 | |
BC068454.1 | - | 1 - 2 | 345 | 159 | |
BI562417.1 | - | - | 350 | 159 | |
BT007367.1 | - | 1 - 2 | 206 | 159 | |
DA652213.1 | - | 1 - 2 | 367 | 159 | |
GQ131806.1 | - | 1 - 2 | 290 | 159 |