The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162429.2 | NP_001155901.1 | 4 - 5 | 621 | 121 |
NM_001256192.1 | NP_001243121.1 | 4 - 5 | 621 | 121 | |
NM_013374.5 | NP_037506.2 | 4 - 5 | 621 | 121 | |
XM_011533252.1 | XP_011531554.1 | 4 - 5 | 844 | 121 | |
XM_011533253.1 | XP_011531555.1 | 4 - 5 | 383 | 121 | |
XM_017005488.1 | XP_016860977.1 | 4 - 5 | 383 | 121 | |
GenBank mRNA | AF151793.1 | - | 4 - 5 | 589 | 121 |
AF250394.1 | - | 4 - 5 | 553 | 121 | |
AF349951.1 | - | 4 - 5 | 594 | 121 | |
BC020066.1 | - | 4 - 5 | 571 | 121 | |
BC068454.1 | - | 4 - 5 | 602 | 121 | |
BT007367.1 | - | 4 - 5 | 463 | 121 | |
GQ131806.1 | - | 4 - 5 | 547 | 121 |