The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256741.1 | NP_001243670.1 | 13 - 14 | 1939 | 66 |
NM_001256742.1 | NP_001243671.1 | 14 - 15 | 1969 | 66 | |
NM_015442.2 | NP_056257.1 | 14 - 15 | 2020 | 66 | |
NR_046352.1 | 12 - 13 | 1777 | 66 | ||
XM_006713084.3 | XP_006713147.1 | 14 - 15 | 1973 | 66 | |
XM_006713085.3 | XP_006713148.1 | 14 - 15 | 1976 | 66 | |
XM_011533566.2 | XP_011531868.1 | 13 - 14 | 1895 | 66 | |
XM_011533567.2 | XP_011531869.1 | 10 - 11 | 1237 | 66 | |
XM_017006109.1 | XP_016861598.1 | 14 - 15 | 2017 | 66 | |
XM_017006110.1 | XP_016861599.1 | 14 - 15 | 2020 | 66 | |
XM_017006111.1 | XP_016861600.1 | 14 - 15 | 2017 | 66 | |
XM_017006112.1 | XP_016861601.1 | 13 - 14 | 1939 | 66 | |
GenBank mRNA | AK022576.1 | - | 14 - 15 | 1822 | 66 |
AK022952.1 | - | 14 - 15 | 1959 | 66 | |
AK023227.1 | - | 14 - 15 | 1977 | 66 | |
AK027026.1 | - | 11 - 12 | 1462 | 66 | |
AK298693.1 | - | 13 - 14 | 1658 | 66 | |
AK302196.1 | - | 14 - 15 | 1961 | 66 | |
AK309670.1 | - | 12 - 13 | 1702 | 66 | |
AL117639.1 | - | 3 - 4 | 271 | 66 | |
BC002928.2 | - | 13 - 14 | 1841 | 66 | |
BC002931.2 | - | 14 - 15 | 1928 | 66 |