The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012109.2 | NP_036241.1 | 6 - 7 | 871 | 75 |
GenBank mRNA | AF186264.1 | - | 6 - 7 | 841 | 75 |
BC010446.1 | - | 6 - 7 | 812 | 75 |