The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052890.3 | NP_443122.3 | 3 - 4 | 1481 | 94 |
XM_006722633.3 | XP_006722696.1 | 3 - 4 | 1478 | 94 | |
GenBank mRNA | AB073610.1 | - | 3 - 4 | 1430 | 94 |
AF384856.1 | - | 3 - 4 | 1374 | 94 | |
AK289415.1 | - | 3 - 4 | 1414 | 94 | |
AK292292.1 | - | 3 - 4 | 1829 | 94 | |
AY358156.1 | - | 3 - 4 | 1405 | 94 | |
BC136551.1 | - | 3 - 4 | 1370 | 94 | |
BC136552.1 | - | 3 - 4 | 1370 | 94 | |
BC144238.1 | - | 3 - 4 | 1370 | 94 |