The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022748.11 | NP_073585.8 | 1 - 2 | 94 | 135 |
GenBank mRNA | AK092864.1 | - | 1 - 2 | 94 | 135 |