Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZDBF2
Entrez Gene ID
57683
Gene Name
zinc finger DBF-type containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 206274641 - 206314438 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285549.1 NP_001272478.1 7 - 7 7453 150
NM_020923.2 NP_065974.1 5 - 5 7560 150
XM_005246711.3 XP_005246768.1 6 - 6 7560 150
XM_005246712.4 XP_005246769.1 6 - 6 7383 150
XM_005246713.3 XP_005246770.1 6 - 6 7377 150
XM_006712656.3 XP_006712719.1 6 - 6 7416 150
XM_011511532.2 XP_011509834.1 6 - 6 7593 150
XM_011511533.2 XP_011509835.1 6 - 6 7410 150
XM_011511534.2 XP_011509836.1 5 - 5 7333 150
XM_011511535.2 XP_011509837.1 5 - 5 7339 150
XM_017004573.1 XP_016860062.1 7 - 7 7666 150
XM_017004574.1 XP_016860063.1 7 - 7 7592 150
XM_017004576.1 XP_016860065.1 2 - 2 5562 150
GenBank mRNA AB046791.2 - 1 - 1 5428 150
AB775780.1 - 7 - 7 7454 150
AB775781.1 - 5 - 5 7560 150

View Full Product Details