The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001137550.1 | NP_001131022.1 | 13 - 14 | 860 | 87 |
NM_001137552.1 | NP_001131024.1 | 6 - 7 | 704 | 87 | |
XM_005246141.4 | XP_005246198.1 | 4 - 5 | 393 | 87 | |
XM_017005253.1 | XP_016860742.1 | 14 - 15 | 922 | 87 | |
XM_017005254.1 | XP_016860743.1 | 14 - 15 | 922 | 87 | |
XM_017005256.1 | XP_016860745.1 | 14 - 15 | 922 | 87 | |
XM_017005258.1 | XP_016860747.1 | 14 - 15 | 922 | 87 | |
GenBank mRNA | AF115510.1 | - | 13 - 14 | 729 | 87 |
BC108914.1 | - | 6 - 7 | 529 | 87 |