The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267803.1 | NP_001254732.1 | 12 - 13 | 1229 | 68 |
NM_001267804.1 | NP_001254733.1 | 12 - 13 | 1220 | 68 | |
NM_001267805.1 | NP_001254734.1 | 12 - 13 | 1216 | 68 | |
NM_001267806.1 | NP_001254735.1 | 11 - 12 | 1103 | 68 | |
NM_025155.2 | NP_079431.1 | 11 - 12 | 1160 | 68 | |
XM_005274308.2 | XP_005274365.2 | 9 - 10 | 1752 | 68 | |
XM_005274309.1 | XP_005274366.1 | 10 - 11 | 1019 | 68 | |
XM_017018357.1 | XP_016873846.1 | 12 - 13 | 1148 | 68 | |
XM_017018358.1 | XP_016873847.1 | 12 - 13 | 1153 | 68 | |
XM_017018359.1 | XP_016873848.1 | 9 - 10 | 1711 | 68 | |
GenBank mRNA | AF087895.1 | - | 12 - 13 | 1154 | 68 |
AK021910.1 | - | 11 - 12 | 1141 | 68 | |
AK222501.1 | - | 11 - 12 | 1141 | 68 | |
AK225520.1 | - | 11 - 12 | 1128 | 68 | |
AK298258.1 | - | 11 - 12 | 1103 | 68 | |
AK309133.1 | - | 9 - 10 | 1367 | 68 | |
AK316394.1 | - | 11 - 12 | 1374 | 68 | |
BC006142.2 | - | 11 - 12 | 1113 | 68 | |
BC021541.1 | - | 11 - 12 | 1124 | 68 | |
BC028628.1 | - | 12 - 13 | 1194 | 68 |