The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267803.1 | NP_001254732.1 | 7 - 8 | 649 | 61 |
NM_001267804.1 | NP_001254733.1 | 7 - 8 | 640 | 61 | |
NM_001267805.1 | NP_001254734.1 | 7 - 8 | 636 | 61 | |
NM_001267806.1 | NP_001254735.1 | 6 - 7 | 523 | 61 | |
NM_025155.2 | NP_079431.1 | 6 - 7 | 580 | 61 | |
XM_005274308.2 | XP_005274365.2 | 4 - 5 | 1172 | 61 | |
XM_005274309.1 | XP_005274366.1 | 5 - 6 | 439 | 61 | |
XM_017018357.1 | XP_016873846.1 | 7 - 8 | 568 | 61 | |
XM_017018358.1 | XP_016873847.1 | 7 - 8 | 573 | 61 | |
XM_017018359.1 | XP_016873848.1 | 4 - 5 | 1131 | 61 | |
GenBank mRNA | AF087895.1 | - | 7 - 8 | 574 | 61 |
AK021910.1 | - | 6 - 7 | 561 | 61 | |
AK222501.1 | - | 6 - 7 | 561 | 61 | |
AK225520.1 | - | 6 - 7 | 548 | 61 | |
AK298258.1 | - | 6 - 7 | 523 | 61 | |
AK309133.1 | - | 4 - 5 | 787 | 61 | |
AK316394.1 | - | 6 - 7 | 794 | 61 | |
BC006142.2 | - | 6 - 7 | 533 | 61 | |
BC021541.1 | - | 6 - 7 | 544 | 61 | |
BC028628.1 | - | 7 - 8 | 614 | 61 | |
CD243380.1 | - | 7 - 8 | 626 | 61 |