The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282786.1 | NP_001269715.1 | 3 - 4 | 408 | 101 |
NM_020200.6 | NP_064585.1 | 3 - 4 | 408 | 101 | |
GenBank mRNA | AB209792.1 | - | 3 - 4 | 358 | 101 |
AF226056.1 | - | 3 - 4 | 356 | 101 | |
AK021950.1 | - | 3 - 4 | 368 | 101 | |
AK222674.1 | - | 3 - 4 | 367 | 101 | |
AK294130.1 | - | 3 - 4 | 367 | 101 | |
AK304055.1 | - | 3 - 4 | 367 | 101 | |
BC008662.2 | - | 3 - 4 | 370 | 101 |