The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007027.3 | NP_008958.2 | 1 - 2 | 123 | 131 |
XM_005247076.3 | XP_005247133.1 | 1 - 2 | 610 | 131 | |
XM_011512357.2 | XP_011510659.1 | 1 - 2 | 194 | 131 | |
GenBank mRNA | AB019397.1 | - | 1 - 2 | 70 | 131 |
AK302584.1 | - | 1 - 2 | 89 | 131 | |
BC032633.1 | - | 1 - 2 | 134 | 131 | |
BC151237.1 | - | 1 - 2 | 76 | 131 | |
D87448.1 | - | 1 - 2 | 76 | 131 |