The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026939.1 | 4 - 5 | 557 | 84 | |
GenBank mRNA | AY226450.1 | - | 4 - 5 | 373 | 84 |
BC112306.1 | - | 3 - 4 | 281 | 84 | |
BC112308.1 | - | 3 - 4 | 281 | 84 |