The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193640.1 | NP_001180569.1 | 1 - 2 | 285 | 142 |
NM_001257966.1 | NP_001244895.1 | 1 - 2 | 285 | 142 | |
NM_201253.2 | NP_957705.1 | 1 - 2 | 285 | 142 | |
NR_047563.1 | 1 - 2 | 285 | 142 | ||
NR_047564.1 | 1 - 2 | 285 | 142 | ||
XM_011509365.2 | XP_011507667.1 | 1 - 2 | 210 | 142 | |
XM_011509367.1 | XP_011507669.1 | 1 - 2 | 210 | 142 | |
XM_017000851.1 | XP_016856340.1 | 1 - 2 | 117 | 142 | |
XM_017000852.1 | XP_016856341.1 | 1 - 2 | 210 | 142 | |
GenBank mRNA | AB208923.1 | - | 1 - 2 | 213 | 142 |
AF154671.1 | - | 1 - 2 | 211 | 142 | |
AK289733.1 | - | 1 - 2 | 230 | 142 | |
AK299368.1 | - | 1 - 2 | 249 | 142 | |
AK307844.1 | - | 1 - 2 | 228 | 142 | |
AY043324.1 | - | 1 - 2 | 211 | 142 | |
AY043325.1 | - | 1 - 2 | 211 | 142 | |
BC136271.1 | - | 1 - 2 | 124 | 142 | |
BC143726.1 | - | 1 - 2 | 124 | 142 | |
DC370545.1 | - | 1 - 2 | 287 | 142 | |
DC370777.1 | - | 1 - 2 | 281 | 142 |