The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018190.3 | NP_060660.2 | 11 - 12 | 1414 | 136 |
NM_176824.2 | NP_789794.1 | 11 - 12 | 1414 | 136 | |
XM_005263106.3 | XP_005263163.1 | 11 - 12 | 1440 | 136 | |
XM_011532079.2 | XP_011530381.1 | 11 - 12 | 1485 | 136 | |
XM_011532080.2 | XP_011530382.1 | 11 - 12 | 1482 | 136 | |
XM_011532081.2 | XP_011530383.1 | 11 - 12 | 1485 | 136 | |
XM_017008357.1 | XP_016863846.1 | 11 - 12 | 1437 | 136 | |
XM_017008358.1 | XP_016863847.1 | 11 - 12 | 1440 | 136 | |
GenBank mRNA | AF521643.1 | - | 11 - 12 | 1378 | 136 |
AF521644.1 | - | 11 - 12 | 1378 | 136 | |
AK001577.1 | - | 11 - 12 | 1378 | 136 | |
AK308076.1 | - | 11 - 12 | 1403 | 136 | |
BC032691.1 | - | 11 - 12 | 1331 | 136 |