The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018190.3 | NP_060660.2 | 15 - 16 | 1856 | 122 |
NM_176824.2 | NP_789794.1 | 15 - 16 | 1856 | 122 | |
XM_005263106.3 | XP_005263163.1 | 15 - 16 | 1882 | 122 | |
XM_011532079.2 | XP_011530381.1 | 15 - 16 | 1927 | 122 | |
XM_011532080.2 | XP_011530382.1 | 15 - 16 | 1924 | 122 | |
GenBank mRNA | AF521643.1 | - | 15 - 16 | 1820 | 122 |
AF521644.1 | - | 15 - 16 | 1820 | 122 | |
AK001577.1 | - | 15 - 16 | 1820 | 122 | |
AK308076.1 | - | 15 - 16 | 1845 | 122 | |
BC032691.1 | - | 15 - 16 | 1773 | 122 |