The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176824.2 | NP_789794.1 | 18 - 19 | 2194 | 157 |
XM_005263106.3 | XP_005263163.1 | 18 - 19 | 2220 | 157 | |
XM_011532079.2 | XP_011530381.1 | 18 - 19 | 2265 | 157 | |
XM_011532080.2 | XP_011530382.1 | 18 - 19 | 2262 | 157 | |
XM_011532081.2 | XP_011530383.1 | 17 - 18 | 2100 | 157 | |
XM_017008357.1 | XP_016863846.1 | 17 - 18 | 2052 | 157 | |
GenBank mRNA | AF521644.1 | - | 18 - 19 | 2158 | 157 |
AK308076.1 | - | 18 - 19 | 2183 | 157 |