The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009922.2 | NP_001009922.1 | 7 - 8 | 774 | 74 |
NM_001278536.1 | NP_001265465.1 | 7 - 8 | 681 | 74 | |
NM_001278537.1 | NP_001265466.1 | 6 - 7 | 654 | 74 | |
NM_001278538.1 | NP_001265467.1 | 8 - 9 | 757 | 74 | |
NM_001278539.1 | NP_001265468.1 | 8 - 9 | 839 | 74 | |
NM_015436.3 | NP_056251.2 | 8 - 9 | 801 | 74 | |
NR_037913.1 | 8 - 9 | 829 | 74 | ||
NR_037914.1 | 8 - 9 | 825 | 74 | ||
NR_103723.1 | 8 - 9 | 799 | 74 | ||
NR_103724.1 | 8 - 9 | 763 | 74 | ||
NR_103725.1 | 6 - 7 | 548 | 74 | ||
XM_011531838.2 | XP_011530140.1 | 8 - 9 | 770 | 74 | |
XM_011531839.2 | XP_011530141.1 | 8 - 9 | 958 | 74 | |
GenBank mRNA | AF247041.1 | - | 8 - 9 | 657 | 74 |
AF255666.1 | - | 8 - 9 | 683 | 74 | |
AF305424.1 | - | 8 - 9 | 691 | 74 | |
AK091501.1 | - | 8 - 9 | 772 | 74 | |
AK308754.1 | - | 7 - 8 | 534 | 74 | |
AK308848.1 | - | 5 - 6 | 583 | 74 | |
AK309555.1 | - | 8 - 9 | 672 | 74 | |
AL050144.1 | - | 7 - 8 | 558 | 74 | |
AY888047.1 | - | 8 - 9 | 645 | 74 | |
BC015464.1 | - | 3 - 4 | 226 | 74 | |
BC047393.1 | - | 8 - 9 | 801 | 74 | |
BG703058.1 | - | 8 - 9 | 670 | 74 | |
BI913695.1 | - | 6 - 7 | 466 | 74 | |
BP314571.1 | - | 6 - 7 | 521 | 74 | |
BQ959587.1 | - | 8 - 9 | 630 | 74 | |
CF597260.1 | - | 6 - 7 | 370 | 74 | |
GQ250944.1 | - | 8 - 9 | 657 | 74 | |
GQ250945.1 | - | 7 - 8 | 630 | 74 | |
GU937000.1 | - | 8 - 9 | 685 | 74 | |
JF691538.1 | - | 8 - 9 | 657 | 74 |