The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136474.2 | NP_001129946.1 | 2 - 3 | 278 | 141 |
GenBank mRNA | AK022567.1 | - | 2 - 3 | 204 | 141 |
DC350527.1 | - | 2 - 3 | 253 | 141 |