The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141980.1 | NP_001135452.1 | 23 - 24 | 5204 | 88 |
NM_005657.2 | NP_005648.1 | 23 - 24 | 5224 | 88 | |
XM_011521985.1 | XP_011520287.1 | 22 - 23 | 5053 | 88 | |
XM_017022533.1 | XP_016878022.1 | 24 - 25 | 5308 | 88 | |
GenBank mRNA | AF078776.1 | - | 23 - 24 | 5249 | 88 |
AK123418.1 | - | 2 - 3 | 1210 | 88 | |
BC112161.1 | - | 23 - 24 | 5184 | 88 | |
BX537418.1 | - | 23 - 24 | 5203 | 88 | |
U09477.1 | - | 10 - 11 | 2243 | 88 |