The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141979.1 | NP_001135451.1 | 2 - 3 | 302 | 70 |
NM_001141980.1 | NP_001135452.1 | 2 - 3 | 302 | 70 | |
NM_005657.2 | NP_005648.1 | 2 - 3 | 322 | 70 | |
XM_011521985.1 | XP_011520287.1 | 2 - 3 | 301 | 70 | |
XM_011521986.2 | XP_011520288.1 | 2 - 3 | 301 | 70 | |
XM_017022533.1 | XP_016878022.1 | 2 - 3 | 257 | 70 | |
GenBank mRNA | AB210025.1 | - | 2 - 3 | 273 | 70 |
AF078776.1 | - | 2 - 3 | 347 | 70 | |
AK292915.1 | - | 2 - 3 | 236 | 70 | |
AK295764.1 | - | 2 - 3 | 302 | 70 | |
AK307405.1 | - | 2 - 3 | 302 | 70 | |
BC112161.1 | - | 2 - 3 | 282 | 70 | |
BP228994.1 | - | 2 - 3 | 302 | 70 | |
BX537418.1 | - | 2 - 3 | 301 | 70 |