The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014306.4 | NP_055121.1 | 10 - 11 | 1420 | 119 |
GenBank mRNA | AF155658.1 | - | 10 - 11 | 1368 | 119 |
AF161466.1 | - | 10 - 11 | 1379 | 119 | |
AK312503.1 | - | 10 - 11 | 1380 | 119 | |
AL050255.1 | - | 10 - 11 | 1364 | 119 | |
AL137272.1 | - | 3 - 4 | 366 | 119 | |
BC000151.1 | - | 10 - 11 | 1358 | 119 | |
BC002970.1 | - | 10 - 11 | 1358 | 119 | |
BC010308.1 | - | 10 - 11 | 1370 | 119 | |
BC016707.1 | - | 10 - 11 | 1385 | 119 | |
CR456450.1 | - | 10 - 11 | 1335 | 119 |