The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014306.4 | NP_055121.1 | 9 - 10 | 1310 | 106 |
GenBank mRNA | AF155658.1 | - | 9 - 10 | 1258 | 106 |
AF161466.1 | - | 9 - 10 | 1269 | 106 | |
AK297826.1 | - | 9 - 10 | 1187 | 106 | |
AK312503.1 | - | 9 - 10 | 1270 | 106 | |
AL050255.1 | - | 9 - 10 | 1254 | 106 | |
AL137272.1 | - | 2 - 3 | 256 | 106 | |
BC000151.1 | - | 9 - 10 | 1248 | 106 | |
BC002970.1 | - | 9 - 10 | 1248 | 106 | |
BC010308.1 | - | 9 - 10 | 1260 | 106 | |
BC016707.1 | - | 9 - 10 | 1275 | 106 | |
CR456450.1 | - | 9 - 10 | 1225 | 106 | |
CR457175.1 | - | 9 - 10 | 1179 | 106 |