Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr.10: 91923769 - 92030998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003972.2 NP_003963.1 14 - 15 1746 95
XM_011540326.1 XP_011538628.1 14 - 15 1845 95
XM_011540327.2 XP_011538629.1 11 - 12 1511 95
XM_011540328.2 XP_011538630.1 11 - 12 1339 95
XM_011540329.2 XP_011538631.1 14 - 15 1956 95
XM_017016877.1 XP_016872366.1 13 - 14 1808 95
XM_017016878.1 XP_016872367.1 13 - 14 1638 95
XM_017016879.1 XP_016872368.1 13 - 14 1762 95
XM_017016880.1 XP_016872369.1 13 - 14 1766 95
XM_017016881.1 XP_016872370.1 12 - 13 1615 95
GenBank mRNA AF038362.1 - 14 - 15 1765 95
AJ001017.2 - 14 - 15 1660 95
BC030273.1 - 8 - 9 993 95
BC047009.2 - 2 - 3 238 95
BC112201.1 - 14 - 15 1730 95

View Full Product Details