The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003972.2 | NP_003963.1 | 17 - 18 | 2143 | 72 |
XM_011540326.1 | XP_011538628.1 | 17 - 18 | 2242 | 72 | |
XM_011540327.2 | XP_011538629.1 | 14 - 15 | 1908 | 72 | |
XM_011540328.2 | XP_011538630.1 | 14 - 15 | 1736 | 72 | |
XM_011540329.2 | XP_011538631.1 | 17 - 18 | 2353 | 72 | |
XM_017016877.1 | XP_016872366.1 | 16 - 17 | 2205 | 72 | |
XM_017016878.1 | XP_016872367.1 | 16 - 17 | 2035 | 72 | |
XM_017016879.1 | XP_016872368.1 | 16 - 17 | 2159 | 72 | |
XM_017016880.1 | XP_016872369.1 | 16 - 17 | 2163 | 72 | |
XM_017016881.1 | XP_016872370.1 | 15 - 16 | 2012 | 72 | |
GenBank mRNA | AB208843.1 | - | 4 - 5 | 432 | 72 |
AF038362.1 | - | 17 - 18 | 2162 | 72 | |
AJ001017.2 | - | 17 - 18 | 2057 | 72 | |
BC030273.1 | - | 11 - 12 | 1390 | 72 | |
BC047009.2 | - | 5 - 6 | 635 | 72 | |
BC112201.1 | - | 17 - 18 | 2127 | 72 |