The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003972.2 | NP_003963.1 | 23 - 24 | 3413 | 96 |
XM_011540326.1 | XP_011538628.1 | 23 - 24 | 3512 | 96 | |
XM_011540327.2 | XP_011538629.1 | 20 - 21 | 3178 | 96 | |
XM_011540328.2 | XP_011538630.1 | 20 - 21 | 3006 | 96 | |
XM_017016877.1 | XP_016872366.1 | 22 - 23 | 3475 | 96 | |
XM_017016878.1 | XP_016872367.1 | 22 - 23 | 3305 | 96 | |
XM_017016879.1 | XP_016872368.1 | 22 - 23 | 3429 | 96 | |
XM_017016880.1 | XP_016872369.1 | 22 - 23 | 3433 | 96 | |
XM_017016881.1 | XP_016872370.1 | 21 - 22 | 3282 | 96 | |
GenBank mRNA | AB208843.1 | - | 10 - 11 | 1702 | 96 |
AF038362.1 | - | 23 - 24 | 3432 | 96 | |
AJ001017.2 | - | 23 - 24 | 3327 | 96 | |
BC112201.1 | - | 23 - 24 | 3397 | 96 |