The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243351.1 | NP_001230280.1 | 2 - 3 | 207 | 154 |
NM_016118.4 | NP_057202.3 | 2 - 3 | 207 | 154 | |
XM_005250011.2 | XP_005250068.1 | 2 - 3 | 272 | 154 | |
XM_017012304.1 | XP_016867793.1 | 2 - 3 | 269 | 154 | |
XM_017012305.1 | XP_016867794.1 | 2 - 3 | 269 | 154 | |
XM_017012306.1 | XP_016867795.1 | 2 - 3 | 288 | 154 | |
XM_017012307.1 | XP_016867796.1 | 2 - 3 | 269 | 154 | |
XM_017012308.1 | XP_016867797.1 | 2 - 3 | 269 | 154 | |
XM_017012309.1 | XP_016867798.1 | 2 - 3 | 272 | 154 | |
XM_017012310.1 | XP_016867799.1 | 2 - 3 | 268 | 154 | |
XM_017012311.1 | XP_016867800.1 | 2 - 3 | 172 | 154 | |
XM_017012312.1 | XP_016867801.1 | 2 - 3 | 144 | 154 | |
XM_017012313.1 | XP_016867802.1 | 2 - 3 | 268 | 154 | |
XM_017012314.1 | XP_016867803.1 | 2 - 3 | 144 | 154 | |
GenBank mRNA | AF155099.1 | - | 2 - 3 | 139 | 154 |
AF300717.1 | - | 2 - 3 | 196 | 154 | |
AF459743.1 | - | 2 - 3 | 155 | 154 | |
AK293159.1 | - | 2 - 3 | 144 | 154 | |
AY129295.1 | - | 2 - 3 | 155 | 154 | |
BC046354.2 | - | 1 - 2 | 123 | 154 | |
DB048887.1 | - | 2 - 3 | 207 | 154 |