Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM33
Entrez Gene ID
155435
Gene Name
RNA binding motif protein 33
Gene Aliases
PRR8
Chromosome Location
Chr.7: 155644494 - 155781485 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053043.2 NP_444271.2 6 - 7 1104 148
XM_005249531.1 XP_005249588.1 7 - 8 1239 148
XM_005249532.1 XP_005249589.1 7 - 8 1239 148
XM_005249533.1 XP_005249590.1 7 - 8 1239 148
XM_005249534.1 XP_005249591.1 6 - 7 1119 148
XM_005249535.1 XP_005249592.1 6 - 7 1119 148
XM_011515840.1 XP_011514142.1 7 - 8 1239 148
GenBank mRNA AK307515.1 - 6 - 7 934 148
AL137724.1 - 4 - 5 601 148

View Full Product Details