The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166222.1 | NP_001159694.1 | 8 - 9 | 1456 | 130 |
NM_020811.1 | NP_065862.1 | 7 - 8 | 1429 | 130 | |
XM_011545189.2 | XP_011543491.1 | 7 - 8 | 1746 | 130 | |
XM_011545191.2 | XP_011543493.1 | 6 - 7 | 2958 | 130 | |
XM_017018054.1 | XP_016873543.1 | 7 - 8 | 2660 | 130 | |
XM_017018055.1 | XP_016873544.1 | 5 - 6 | 1940 | 130 | |
GenBank mRNA | AB037815.1 | - | 6 - 7 | 2554 | 130 |
AK294035.1 | - | 9 - 10 | 130 | ||
EF560746.1 | - | 7 - 8 | 1434 | 130 |